Product Details

SNP ID
rs3813732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7109961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCATGTGCGCGCGCACACACA[C/G]CTCTCAGAGAAGAATCTATTGCTTC
Phenotype
MIM: 608974 MIM: 611866
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C1RL PubMed Links

Gene Details

Gene
C1RL
Gene Name
complement C1r subcomponent like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297640.1 Intron NP_001284569.1
NM_001297642.1 Intron NP_001284571.1
NM_001297643.1 Intron NP_001284572.1
NM_016546.3 Intron NP_057630.2
XM_017019397.1 Intron XP_016874886.1
XM_017019398.1 Intron XP_016874887.1
XM_017019399.1 Intron XP_016874888.1
XM_017019400.1 Intron XP_016874889.1
XM_017019401.1 Intron XP_016874890.1
XM_017019402.1 Intron XP_016874891.1
XM_017019403.1 Intron XP_016874892.1
XM_017019404.1 Intron XP_016874893.1
Gene
C1RL-AS1
Gene Name
C1RL antisense RNA 1
There are no transcripts associated with this gene.

Gene
RBP5
Gene Name
retinol binding protein 5
There are no transcripts associated with this gene.

View Full Product Details