Product Details

SNP ID
rs1562855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38781326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCATAACTTGTTTAATCCTTTAA[C/T]TGTATCCCACAATGATTGGCTGTAG
Phenotype
MIM: 607006
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GEMIN6 PubMed Links

Gene Details

Gene
GEMIN6
Gene Name
gem nuclear organelle associated protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024775.9 Intron NP_079051.9

View Full Product Details