Product Details

SNP ID
rs10206639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:106097378 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCCAGGCCCCACTGGCCAGCAGTA[C/T]ACAGCCCCACCAGTGACTGTGGGCC
Phenotype
MIM: 609749
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UXS1 PubMed Links

Gene Details

Gene
UXS1
Gene Name
UDP-glucuronate decarboxylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253875.1 Intron NP_001240804.1
NM_001253876.1 Intron NP_001240805.1
NM_025076.4 Intron NP_079352.2
XM_006712775.3 Intron XP_006712838.1
XM_011511902.2 Intron XP_011510204.2
XM_011511903.2 Intron XP_011510205.2
XM_011511904.2 Intron XP_011510206.1
XM_017005008.1 Intron XP_016860497.1
XM_017005009.1 Intron XP_016860498.1
XM_017005010.1 Intron XP_016860499.1
XM_017005011.1 Intron XP_016860500.1
XM_017005012.1 Intron XP_016860501.1
XM_017005013.1 Intron XP_016860502.1
XM_017005014.1 Intron XP_016860503.1

View Full Product Details