Product Details

SNP ID
rs6478973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99106999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAGGTAACATTTCCACACCGCT[A/G]TTAACTTGAATAACACAGTGCACTC
Phenotype
MIM: 190181
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGFBR1 PubMed Links

Gene Details

Gene
TGFBR1
Gene Name
transforming growth factor beta receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130916.2 Intron NP_001124388.1
NM_001306210.1 Intron NP_001293139.1
NM_004612.3 Intron NP_004603.1
XM_011518948.2 Intron XP_011517250.1
XM_011518949.2 Intron XP_011517251.1
XM_011518950.2 Intron XP_011517252.1
XM_017015063.1 Intron XP_016870552.1

View Full Product Details