Product Details

SNP ID
rs1226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:41127707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACGATATATTACTATTTAGTCTAAG[C/T]TTTAATTCAAAGGTTGAGAATGACG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KBTBD6 PubMed Links

Gene Details

Gene
KBTBD6
Gene Name
kelch repeat and BTB domain containing 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152903.4 5094 UTR 3 NP_690867.3
Gene
LOC101929140
Gene Name
uncharacterized LOC101929140
There are no transcripts associated with this gene.

View Full Product Details