Product Details

SNP ID
rs9695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94603360 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTCCAGGTAGAGGCAATTCTCC[A/G]GATGCAGGCAGGGCAGGTGCTCACT
Phenotype
MIM: 611570 MIM: 603027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBP1 PubMed Links

Gene Details

Gene
FBP1
Gene Name
fructose-bisphosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000507.3 1369 UTR 3 NP_000498.2
NM_001127628.1 1369 UTR 3 NP_001121100.1
XM_006717005.3 1369 UTR 3 XP_006717068.1
Gene
FBP2
Gene Name
fructose-bisphosphatase 2
There are no transcripts associated with this gene.

View Full Product Details