Product Details
- SNP ID
-
rs3805167
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:177431892 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATACAGATACTGCTTATTTGACTA[C/G]ACACTGGCTACTGTATACCTTATGT
- Phenotype
-
MIM: 613228
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AGA
PubMed Links
Gene Details
- Gene
- AGA
- Gene Name
- aspartylglucosaminidase
View Full Product Details