Product Details

SNP ID
rs11247571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1005262 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGGGAACTGAAAAGCAGTAGAAG[A/G]AACAGTCAGAGATGCCTCACTGATA
Phenotype
MIM: 600365 MIM: 607251
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABR PubMed Links

Gene Details

Gene
ABR
Gene Name
active BCR-related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001092.4 3600 UTR 3 NP_001083.2
NM_001159746.2 3600 UTR 3 NP_001153218.1
NM_001256847.2 3600 UTR 3 NP_001243776.1
NM_001282149.1 3600 UTR 3 NP_001269078.1
NM_001322840.1 3600 UTR 3 NP_001309769.1
NM_001322841.1 3600 UTR 3 NP_001309770.1
NM_001322842.1 3600 Intron NP_001309771.1
NM_021962.4 3600 UTR 3 NP_068781.2
XM_011523810.2 3600 UTR 3 XP_011522112.1
XM_011523811.2 3600 Intron XP_011522113.2
XM_011523812.2 3600 UTR 3 XP_011522114.1
XM_011523813.2 3600 UTR 3 XP_011522115.1
XM_011523814.2 3600 UTR 3 XP_011522116.1
XM_011523815.2 3600 UTR 3 XP_011522117.1
XM_017024539.1 3600 UTR 3 XP_016880028.1
XM_017024540.1 3600 UTR 3 XP_016880029.1
XM_017024541.1 3600 Intron XP_016880030.1
XM_017024542.1 3600 UTR 3 XP_016880031.1
XM_017024543.1 3600 UTR 3 XP_016880032.1
Gene
TIMM22
Gene Name
translocase of inner mitochondrial membrane 22 homolog (yeast)
There are no transcripts associated with this gene.

View Full Product Details