Product Details

SNP ID
rs12461400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16203113 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGGGAGAGCAAGGTGCGTTGGCC[C/T]GGCAAAGGCTCTGAGAAGGTCTGCA
Phenotype
MIM: 603535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP1M1 PubMed Links

Gene Details

Gene
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130524.1 Intron NP_001123996.1
NM_032493.3 Intron NP_115882.1
XM_017027420.1 Intron XP_016882909.1
XM_017027421.1 Intron XP_016882910.1

View Full Product Details