Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000578.3 | Intron | NP_000569.3 | |||
XM_005246793.3 | Intron | XP_005246850.1 | |||
XM_005246794.3 | Intron | XP_005246851.1 | |||
XM_006712709.3 | Intron | XP_006712772.1 | |||
XM_006712710.3 | Intron | XP_006712773.1 | |||
XM_006712711.3 | Intron | XP_006712774.1 | |||
XM_011511684.2 | Intron | XP_011509986.1 | |||
XM_011511685.2 | Intron | XP_011509987.1 | |||
XM_017004765.1 | Intron | XP_016860254.1 | |||
XM_017004766.1 | Intron | XP_016860255.1 | |||
XM_017004767.1 | Intron | XP_016860256.1 |