Product Details

SNP ID
rs7151016
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:32333088 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCACAGGGCTCATTAGCCTTTTCT[A/G]GGTTCCAGGAAGAAGTTAACCTTTG
Phenotype
MIM: 604691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKAP6 PubMed Links

Gene Details

Gene
AKAP6
Gene Name
A-kinase anchoring protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004274.4 Intron NP_004265.3
XM_011537378.2 Intron XP_011535680.1
XM_011537379.2 Intron XP_011535681.1
XM_011537383.2 Intron XP_011535685.1
XM_017021808.1 Intron XP_016877297.1
XM_017021809.1 Intron XP_016877298.1
XM_017021810.1 Intron XP_016877299.1
XM_017021811.1 Intron XP_016877300.1

View Full Product Details