Product Details

SNP ID
rs1522718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63704107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAAGTTCTGCTTCCCTGAAATGG[A/G]CTAAGCAACATTAATTAGACTAGCT
Phenotype
MIM: 615682
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB11 PubMed Links

Gene Details

Gene
SERPINB11
Gene Name
serpin family B member 11 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291278.1 Intron NP_001278207.1
NM_001291279.1 Intron NP_001278208.1
NM_080475.3 Intron NP_536723.2
XM_011526251.1 Intron XP_011524553.1

View Full Product Details