Product Details

SNP ID
rs872251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97995061 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAATGCATGTTGGGTGCTCAAT[A/G]AGTAGTAGTTAGGAAGAATAATTAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANP32B PubMed Links

Gene Details

Gene
ANP32B
Gene Name
acidic nuclear phosphoprotein 32 family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006401.2 Intron NP_006392.1

View Full Product Details