Product Details

SNP ID
rs872298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:134330291 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCTCTGCCAGGCTTGAGTTTGT[A/G]TTTTTTAGCTGGCTTTGGTGGCCAG
Phenotype
MIM: 180245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RXRA PubMed Links

Gene Details

Gene
RXRA
Gene Name
retinoid X receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291920.1 Intron NP_001278849.1
NM_001291921.1 Intron NP_001278850.1
NM_002957.5 Intron NP_002948.1

View Full Product Details