Product Details

SNP ID
rs614507
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.20:58239836 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACATACTGATCTAGCAGCGGCTGAG[G/T]TATTACATCCTGGGAAATGAGAGAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP4R1L PubMed Links

Gene Details

Gene
PPP4R1L
Gene Name
protein phosphatase 4 regulatory subunit 1 like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details