Product Details

SNP ID
rs10902717
Assay Type
Validated
NCBI dbSNP Submissions
47
Location
Chr.1:26159601 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTTGGTTCCCAGGCCTCAACTACC[C/T]ATGAGCAGACTGACTACACACTAGA
Phenotype
MIM: 616698
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM110D PubMed Links

Gene Details

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024869.2 Intron NP_079145.2
Gene
ZNF593
Gene Name
zinc finger protein 593
There are no transcripts associated with this gene.

View Full Product Details