Product Details

SNP ID
rs12824474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10450326 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGAATACTTTCAACAATTATTCTT[C/T]CATTTATTCTTTGACAAATTCAGAA
Phenotype
MIM: 161555
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC1 PubMed Links

Gene Details

Gene
KLRC1
Gene Name
killer cell lectin like receptor C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304448.1 Intron NP_001291377.1
NM_002259.4 Intron NP_002250.1
NM_007328.3 Intron NP_015567.1
NM_213657.2 Intron NP_998822.1
NM_213658.2 Intron NP_998823.1

View Full Product Details