Product Details

SNP ID
rs33935503
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:206109902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTAGGCCTGCCTAGATCTGGG[T/A]CACCATGTGTGCATGGACACCCTAT
Phenotype
MIM: 600264 MIM: 616088
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
AVPR1B PubMed Links

Gene Details

Gene
AVPR1B
Gene Name
arginine vasopressin receptor 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000707.3 1720 UTR 3 NP_000698.1
Gene
C1orf186
Gene Name
chromosome 1 open reading frame 186
There are no transcripts associated with this gene.

View Full Product Details