Product Details

SNP ID
rs11812979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:3072859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAAACATCAGGATACCAGATT[A/G]CACGTTCACAATGGCATTTCGGCTA
Phenotype
MIM: 171840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927824 PubMed Links

Gene Details

Gene
LOC101927824
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

Gene
PFKP
Gene Name
phosphofructokinase, platelet
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242339.1 Intron NP_001229268.1
NM_001323067.1 Intron NP_001309996.1
NM_001323068.1 Intron NP_001309997.1
NM_001323069.1 Intron NP_001309998.1
NM_001323070.1 Intron NP_001309999.1
NM_001323071.1 Intron NP_001310000.1
NM_001323072.1 Intron NP_001310001.1
NM_001323073.1 Intron NP_001310002.1
NM_001323074.1 Intron NP_001310003.1
NM_002627.4 Intron NP_002618.1
XM_005252465.4 Intron XP_005252522.1
XM_005252466.4 Intron XP_005252523.1
XM_005252467.2 Intron XP_005252524.1
XM_006717449.1 Intron XP_006717512.1

View Full Product Details