Product Details

SNP ID
rs9507510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25378985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTCCCATTTAAAAATTCTATAT[A/G]CATATTTGTATACTTTTCTTTTTAA
Phenotype
MIM: 605870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP8A2 PubMed Links

Gene Details

Gene
ATP8A2
Gene Name
ATPase phospholipid transporting 8A2
There are no transcripts associated with this gene.

View Full Product Details