Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152450.2 | Intron | NP_689663.2 | |||
XM_005254166.2 | Intron | XP_005254223.1 | |||
XM_006720398.3 | Intron | XP_006720461.1 | |||
XM_006720399.2 | Intron | XP_006720462.1 | |||
XM_011521247.2 | Intron | XP_011519549.1 | |||
XM_011521248.2 | Intron | XP_011519550.1 | |||
XM_011521249.2 | Intron | XP_011519551.1 | |||
XM_011521250.2 | Intron | XP_011519552.1 | |||
XM_017021931.1 | Intron | XP_016877420.1 | |||
XM_017021932.1 | Intron | XP_016877421.1 |