Product Details

SNP ID
rs4775163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59403283 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCAATATGACTGTATTTGGAGATC[C/T]GGCATTTAAAGAGGAAATTAAGGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM81A PubMed Links

Gene Details

Gene
FAM81A
Gene Name
family with sequence similarity 81 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152450.2 Intron NP_689663.2
XM_005254166.2 Intron XP_005254223.1
XM_006720398.3 Intron XP_006720461.1
XM_006720399.2 Intron XP_006720462.1
XM_011521247.2 Intron XP_011519549.1
XM_011521248.2 Intron XP_011519550.1
XM_011521249.2 Intron XP_011519551.1
XM_011521250.2 Intron XP_011519552.1
XM_017021931.1 Intron XP_016877420.1
XM_017021932.1 Intron XP_016877421.1

View Full Product Details