Product Details
- SNP ID
-
rs11211460
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
48
- Location
-
Chr.1:38843435 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATATCTGAACTATTCCATTAAAAA[C/T]TCAGGCCGGGCGCAGTGGCTCACTC
- Phenotype
-
MIM: 608267
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RRAGC
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115651987] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RRAGC
- Gene Name
- Ras related GTP binding C
View Full Product Details