Product Details

SNP ID
rs12882501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93719809 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATCTAAACCCCAAACAAGTCCTC[A/C]AATAGAAGGAAATGAGGAAAGAAGA
Phenotype
MIM: 613851 MIM: 616884
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRIMA1 PubMed Links

Gene Details

Gene
PRIMA1
Gene Name
proline rich membrane anchor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178013.3 2139 UTR 3 NP_821092.1
XM_011536456.2 2139 UTR 3 XP_011534758.1
Gene
UNC79
Gene Name
unc-79 homolog (C. elegans)
There are no transcripts associated with this gene.

View Full Product Details