Product Details

SNP ID
rs1559162
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163507507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATACGTTTAGAGTAACTTAAACTG[A/T]TATTAGATCTACTTTCCCCCATATA
Phenotype
MIM: 605527
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MAT2B PubMed Links

Gene Details

Gene
MAT2B
Gene Name
methionine adenosyltransferase 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013283.4 Intron NP_037415.1
NM_182796.2 Intron NP_877725.1

View Full Product Details