Product Details
- SNP ID
-
rs10763703
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29296551 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATACTGGGACTTTTTCCTCTAGAC[A/G]AGTAAAGTAGAAAACCTCTCAACTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL1
PubMed Links
Gene Details
- Gene
- LYZL1
- Gene Name
- lysozyme like 1
View Full Product Details