Product Details
- SNP ID
-
rs7100574
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:29295362 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATTTATAAGTACCTCTCATCTC[C/T]GGTTTCTTATCTTCTGAGCTAAAAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LYZL1
PubMed Links
Gene Details
- Gene
- LYZL1
- Gene Name
- lysozyme like 1
View Full Product Details