Product Details

SNP ID
rs3739609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36169601 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGAGTACTGGGACATGGCCCTGAG[C/T]GTGGACAACCACGTCTTCTTTGCAC
Phenotype
MIM: 603960 MIM: 607141
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCIN PubMed Links

Gene Details

Gene
CCIN
Gene Name
calicin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005893.2 210 Silent Mutation AGC,AGT S33S NP_005884.2
Gene
GLIPR2
Gene Name
GLI pathogenesis related 2
There are no transcripts associated with this gene.

View Full Product Details