Product Details

SNP ID
rs12737576
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:151299175 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCAGCCTCTCTCCTTGGTAGCATT[G/T]AATTCTAAGCCATCAAATGAGTAAG
Phenotype
MIM: 602758 MIM: 610568
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PI4KB PubMed Links

Gene Details

Gene
PI4KB
Gene Name
phosphatidylinositol 4-kinase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198774.1 Intron NP_001185703.1
XM_005245264.3 Intron XP_005245321.1
XM_011509634.1 Intron XP_011507936.1
XM_011509635.1 Intron XP_011507937.1
XM_011509636.2 Intron XP_011507938.1
XM_011509637.2 Intron XP_011507939.1
XM_011509638.1 Intron XP_011507940.1
XM_017001486.1 Intron XP_016856975.1
XM_017001487.1 Intron XP_016856976.1
XM_017001488.1 Intron XP_016856977.1
XM_017001489.1 Intron XP_016856978.1
XM_017001490.1 Intron XP_016856979.1
XM_017001491.1 Intron XP_016856980.1
XM_017001492.1 Intron XP_016856981.1
Gene
ZNF687
Gene Name
zinc finger protein 687
There are no transcripts associated with this gene.

View Full Product Details