Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_153038.1 | Intron | NP_694583.1 | |||
XM_017003466.1 | Intron | XP_016858955.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000438.5 | Intron | NP_000429.2 | |||
NM_001127366.2 | Intron | NP_001120838.1 | |||
NM_013942.4 | Intron | NP_039230.1 | |||
NM_181457.3 | Intron | NP_852122.1 | |||
NM_181458.3 | Intron | NP_852123.1 | |||
NM_181459.3 | Intron | NP_852124.1 | |||
NM_181460.3 | Intron | NP_852125.1 | |||
NM_181461.3 | Intron | NP_852126.1 |