Product Details

SNP ID
rs11603869
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:6519995 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAGGCAGAAACCCTTTCTCTCATCA[C/G]AGCTGGTCTTTGATGATCATGGTCA
Phenotype
MIM: 615818
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DNHD1 PubMed Links

Gene Details

Gene
DNHD1
Gene Name
dynein heavy chain domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144666.2 2242 Missense Mutation CAG,GAG Q560E NP_653267.2
NM_173589.3 2242 Missense Mutation CAG,GAG Q560E NP_775860.3
Gene
RRP8
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015324.3 2242 Intron NP_056139.1
XM_011519955.2 2242 Intron XP_011518257.1
XM_017017423.1 2242 Intron XP_016872912.1

View Full Product Details