Product Details

SNP ID
rs4705134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:147241977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATTCAGCCAGGCTTTGTTCTTG[C/T]GGGATGCCCAAGAAAGCTACATAAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STK32A PubMed Links

Gene Details

Gene
STK32A
Gene Name
serine/threonine kinase 32A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001112724.1 Intron NP_001106195.1
NM_001287740.1 Intron NP_001274669.1
NM_145001.3 Intron NP_659438.1
XM_011537576.1 Intron XP_011535878.1
XM_011537577.2 Intron XP_011535879.1
XM_011537579.2 Intron XP_011535881.1
XM_017009213.1 Intron XP_016864702.1
XM_017009214.1 Intron XP_016864703.1

View Full Product Details