Product Details

SNP ID
rs13045798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17700803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACTCAAGGCCTTCTATAACAAA[C/G]AGTAGGGACCCGTTCTGGGCCCCAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BANF2 PubMed Links

Gene Details

Gene
BANF2
Gene Name
barrier to autointegration factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014977.3 Intron NP_001014977.2
NM_001159495.1 Intron NP_001152967.1
NM_178477.4 Intron NP_848572.3
XM_005260668.3 Intron XP_005260725.1
XM_011529170.1 Intron XP_011527472.1
XM_011529171.2 Intron XP_011527473.1
Gene
LOC105372547
Gene Name
uncharacterized LOC105372547
There are no transcripts associated with this gene.

View Full Product Details