Product Details

SNP ID
rs11679767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:189681574 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGCTGTCTGCTTTCTACACCTAC[A/G]TTTTCAGGGTTGATACTCAAAACAC
Phenotype
MIM: 609803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKAR PubMed Links

Gene Details

Gene
ANKAR
Gene Name
ankyrin and armadillo repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144708.3 Intron NP_653309.3
XM_011510673.2 Intron XP_011508975.1
XM_011510674.1 Intron XP_011508976.1
XM_011510675.2 Intron XP_011508977.1
XM_011510676.2 Intron XP_011508978.1
XM_011510677.2 Intron XP_011508979.1
XM_011510679.2 Intron XP_011508981.1
XM_011510680.1 Intron XP_011508982.1
XM_011510681.1 Intron XP_011508983.1
XM_011510682.2 Intron XP_011508984.1
XM_011510685.2 Intron XP_011508987.1
XM_011510686.2 Intron XP_011508988.1
XM_011510687.2 Intron XP_011508989.1
XM_011510688.1 Intron XP_011508990.1
XM_011510689.2 Intron XP_011508991.1
XM_011510691.2 Intron XP_011508993.1
XM_011510692.2 Intron XP_011508994.1
XM_011510693.2 Intron XP_011508995.1
XM_011510694.2 Intron XP_011508996.1
XM_017003413.1 Intron XP_016858902.1
XM_017003414.1 Intron XP_016858903.1
XM_017003415.1 Intron XP_016858904.1
XM_017003416.1 Intron XP_016858905.1
XM_017003417.1 Intron XP_016858906.1
XM_017003418.1 Intron XP_016858907.1
XM_017003419.1 Intron XP_016858908.1
XM_017003420.1 Intron XP_016858909.1

View Full Product Details