Product Details

SNP ID
rs35253356
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:144295352 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTAACAAGATAAGGAAGTATGAAAA[A/G]TAATCTCGACATCTGAGATGGTGAG
Phenotype
MIM: 610596 MIM: 140580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BOP1 PubMed Links

Gene Details

Gene
BOP1
Gene Name
block of proliferation 1
There are no transcripts associated with this gene.

Gene
HSF1
Gene Name
heat shock transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005272315.2 Intron XP_005272372.1
XM_005272316.2 Intron XP_005272373.1
XM_005272317.1 Intron XP_005272374.1
XM_011517004.2 Intron XP_011515306.1
XM_011517006.2 Intron XP_011515308.1
XM_017013377.1 Intron XP_016868866.1

View Full Product Details