Product Details

SNP ID
rs10885650
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:115096314 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCTGTGTTTTTATTATGTATACTC[C/T]TGGGGATAAAGAACACATTGGTATT
Phenotype
MIM: 612869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATRNL1 PubMed Links

Gene Details

Gene
ATRNL1
Gene Name
attractin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276282.3 2051 Intron NP_001263211.1
NM_207303.4 2051 Intron NP_997186.1
XM_011539587.1 2051 Intron XP_011537889.1
XM_011539588.2 2051 Intron XP_011537890.1
XM_011539589.2 2051 Intron XP_011537891.1
XM_011539590.2 2051 UTR 5 XP_011537892.1
XM_011539591.2 2051 Intron XP_011537893.1
XM_017016035.1 2051 Intron XP_016871524.1
XM_017016036.1 2051 Intron XP_016871525.1
XM_017016037.1 2051 Intron XP_016871526.1
XM_017016038.1 2051 Intron XP_016871527.1
XM_017016039.1 2051 Intron XP_016871528.1
XM_017016040.1 2051 Intron XP_016871529.1

View Full Product Details