Product Details

SNP ID
rs111033624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:124346737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTGGATGGCAGCTATTTGCTGA[C/G]GGACAGCGAGAGCGTGCCAGGCGTG
Phenotype
MIM: 300490
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SH2D1A PubMed Links

Gene Details

Gene
SH2D1A
Gene Name
SH2 domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114937.2 456 Missense Mutation ACG,AGG T32R NP_001108409.1
NM_002351.4 456 Missense Mutation ACG,AGG T32R NP_002342.1

View Full Product Details