Product Details

SNP ID
rs137058
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42574674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCAGAGACCGCTGCAGGCCCTG[C/T]CTCACCTCATGTTCCTCCTGGCCCT
Phenotype
MIM: 611520
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLDIP3 PubMed Links

Gene Details

Gene
POLDIP3
Gene Name
DNA polymerase delta interacting protein 3
There are no transcripts associated with this gene.

Gene
RRP7BP
Gene Name
ribosomal RNA processing 7 homolog B, pseudogene
There are no transcripts associated with this gene.

Gene
SERHL2
Gene Name
serine hydrolase-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284334.1 Intron NP_001271263.1
NM_014509.4 Intron NP_055324.2
XM_017028738.1 Intron XP_016884227.1
XM_017028739.1 Intron XP_016884228.1

View Full Product Details