Product Details

SNP ID
rs104664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:45315973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAACTCACCAGAAACCTATCAGCT[A/G]GATCGCAGATGGTTCATTTGTGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM118A PubMed Links

Gene Details

Gene
FAM118A
Gene Name
family with sequence similarity 118 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104595.1 Intron NP_001098065.1
NM_017911.2 Intron NP_060381.2
XM_011530247.2 Intron XP_011528549.1
XM_011530254.2 Intron XP_011528556.1
XM_011530256.1 Intron XP_011528558.1
XM_017028841.1 Intron XP_016884330.1
XM_017028842.1 Intron XP_016884331.1
XM_017028843.1 Intron XP_016884332.1
XM_017028844.1 Intron XP_016884333.1
XM_017028845.1 Intron XP_016884334.1
XM_017028846.1 Intron XP_016884335.1
XM_017028847.1 Intron XP_016884336.1
XM_017028848.1 Intron XP_016884337.1
XM_017028849.1 Intron XP_016884338.1
XM_017028850.1 Intron XP_016884339.1
XM_017028851.1 Intron XP_016884340.1
XM_017028852.1 Intron XP_016884341.1
XM_017028853.1 Intron XP_016884342.1
XM_017028854.1 Intron XP_016884343.1
XM_017028855.1 Intron XP_016884344.1
XM_017028856.1 Intron XP_016884345.1
XM_017028857.1 Intron XP_016884346.1
XM_017028858.1 Intron XP_016884347.1
XM_017028859.1 Intron XP_016884348.1
XM_017028860.1 Intron XP_016884349.1
XM_017028861.1 Intron XP_016884350.1

View Full Product Details