Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001315501.1 | 974 | Intron | NP_001302430.1 | ||
NM_001315502.1 | 974 | Intron | NP_001302431.1 | ||
NM_002555.5 | 974 | Intron | NP_002546.3 | ||
NM_183233.2 | 974 | Intron | NP_899056.2 | ||
XM_011520141.2 | 974 | Intron | XP_011518443.2 | ||
XM_011520142.2 | 974 | Intron | XP_011518444.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001302862.1 | 974 | Intron | NP_001289791.1 | ||
NM_007105.3 | 974 | Intron | NP_009036.2 | ||
XM_017017832.1 | 974 | UTR 5 | XP_016873321.1 | ||
XM_017017833.1 | 974 | UTR 5 | XP_016873322.1 | ||
XM_017017834.1 | 974 | UTR 5 | XP_016873323.1 | ||
XM_017017835.1 | 974 | Intron | XP_016873324.1 | ||
XM_017017836.1 | 974 | Intron | XP_016873325.1 |