Product Details
- SNP ID
-
rs740526
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:74313336 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATAAAATTAGAATGATAAAGCGAC[C/T]CACATCTAATGGCCATCCTAAGAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VRTN
PubMed Links
Gene Details
- Gene
- VRTN
- Gene Name
- vertebrae development associated
View Full Product Details