Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005173.3 | Intron | NP_005164.2 | |||
NM_174953.2 | Intron | NP_777613.1 | |||
NM_174954.2 | Intron | NP_777614.1 | |||
NM_174955.2 | Intron | NP_777615.1 | |||
NM_174956.2 | Intron | NP_777616.1 | |||
NM_174957.2 | Intron | NP_777617.1 | |||
NM_174958.2 | Intron | NP_777618.1 | |||
XM_011523881.2 | Intron | XP_011522183.1 | |||
XM_011523882.2 | Intron | XP_011522184.1 | |||
XM_011523884.2 | Intron | XP_011522186.1 | |||
XM_011523885.1 | Intron | XP_011522187.1 | |||
XM_011523888.2 | Intron | XP_011522190.1 | |||
XM_011523889.1 | Intron | XP_011522191.1 | |||
XM_011523892.2 | Intron | XP_011522194.1 | |||
XM_017024692.1 | Intron | XP_016880181.1 | |||
XM_017024693.1 | Intron | XP_016880182.1 |