Product Details

SNP ID
rs41413
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72178704 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATCCTGCCCTAGAAAAAATTGCA[A/G]TAAAAGCCTGGGGTTAGATCAGGCT
Phenotype
MIM: 607880
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC6B PubMed Links

Gene Details

Gene
EXOC6B
Gene Name
exocyst complex component 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321729.1 2503 UTR 3 NP_001308658.1
NM_001321730.1 2503 UTR 3 NP_001308659.1
NM_001321731.1 2503 UTR 3 NP_001308660.1
NM_001321733.1 2503 UTR 3 NP_001308662.1
NM_001321734.1 2503 UTR 3 NP_001308663.1
NM_015189.2 2503 UTR 3 NP_056004.1
XM_005264224.1 2503 UTR 3 XP_005264281.1
XM_011532711.2 2503 Intron XP_011531013.1
XM_011532712.2 2503 Intron XP_011531014.1
XM_017003641.1 2503 UTR 3 XP_016859130.1
XM_017003642.1 2503 UTR 3 XP_016859131.1

View Full Product Details