Product Details

SNP ID
rs743996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:43595984 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTCCAAATCACAAGCCAAAA[G/T]ATTATTCTAATTATACACATGTAGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IFT52 PubMed Links

Gene Details

Gene
IFT52
Gene Name
intraflagellar transport 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303458.2 Intron NP_001290387.1
NM_001303459.2 Intron NP_001290388.1
NM_001323578.1 Intron NP_001310507.1
NM_001323579.1 Intron NP_001310508.1
NM_001323580.1 Intron NP_001310509.1
NM_001323581.1 Intron NP_001310510.1
NM_016004.4 Intron NP_057088.2
XM_017027863.1 Intron XP_016883352.1

View Full Product Details