Product Details
- SNP ID
-
rs2470612
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:51941557 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGATACCTCTCCTTTCCACTGTCT[C/T]CATGTTTCTTTCAGCTTGGGTCTTT
- Phenotype
-
MIM: 610507
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LEO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2570267] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LEO1
- Gene Name
- LEO1 homolog, Paf1/RNA polymerase II complex component
View Full Product Details