Product Details

SNP ID
rs4816481
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:34364922 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTAGGGGATCATTCTGTAGCCTGGA[A/G]ACAGGGCACAACTAATGACATGTGT
Phenotype
MIM: 603796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE2 PubMed Links

Gene Details

Gene
KCNE2
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172201.1 Intron NP_751951.1

View Full Product Details