Product Details

SNP ID
rs3757062
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:153012004 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATTTAGTTTACAGATGTCTTTGCT[A/T]ACTTGGTTTTATGGTTTCTATGCAT
Phenotype
MIM: 613542 MIM: 607191
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MTRF1L PubMed Links

Gene Details

Gene
MTRF1L
Gene Name
mitochondrial translational release factor 1 like
There are no transcripts associated with this gene.

Gene
RGS17
Gene Name
regulator of G-protein signaling 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012419.4 Intron NP_036551.3
XM_017010733.1 Intron XP_016866222.1

View Full Product Details