Product Details
- SNP ID
-
rs35393404
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41380508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATACCCAGATACACCCTGCCCTG[A/C]CACTTCTGTGTCTTTGTTCAACCTC
- Phenotype
-
MIM: 602763
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT34
PubMed Links
Gene Details
- Gene
- KRT34
- Gene Name
- keratin 34
View Full Product Details