Product Details
- SNP ID
-
rs3796917
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:105557092 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AATTACCTAAATACAAGAAATTTAG[A/T]GGTTGAGTTGTAAAAACTCAGAAAG
- Phenotype
-
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF38
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs6823261] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARHGEF38
- Gene Name
- Rho guanine nucleotide exchange factor 38
- Gene
- ARHGEF38-IT1
- Gene Name
- ARHGEF38 intronic transcript 1
There are no transcripts associated with this gene.
View Full Product Details