Product Details
- SNP ID
-
rs5749132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:30609031 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGCCTTCCTCCTCGGTGTAGTAC[A/G]AGGAAGGATCCCATGACACCTGCTC
- Phenotype
-
MIM: 605819
MIM: 613441
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PES1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117324970] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PES1
- Gene Name
- pescadillo ribosomal biogenesis factor 1
There are no transcripts associated with this gene.
- Gene
- TCN2
- Gene Name
- transcobalamin 2
View Full Product Details